Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081143.1 | NP_001074612.1 | 3 | 304 | ||
GenBank mRNA | AK040065.1 | - | 3 | 305 | ||
BC147449.1 | - | 3 | 325 | 9056091 | ||
BC147466.1 | - | 3 | 325 | 9056108 | ||
BC147839.1 | - | 3 | 253 | 9087556 | ||
BC147841.1 | - | 3 | 253 | 9087558 |