Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175520.4 | NP_780729.2 | 1 | 697 | ||
GenBank mRNA | AK029064.1 | - | 1 | 682 | ||
AK046700.1 | - | 1 | 698 | |||
BC141305.1 | - | 1 | 568 | 9055917 | ||
EU809459.1 | - | 1 | 303 | |||
EU809460.1 | - | 1 | 279 |