Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008928.4 | NP_032954.1 | 9 | 880 | ||
GenBank mRNA | AK008141.1 | - | 9 | 883 | ||
AK011002.1 | - | 9 | 881 | |||
AK053378.1 | - | 9 | 881 | |||
AK150067.1 | - | 9 | 881 | |||
AK159379.1 | - | 9 | 879 | |||
AK166255.1 | - | 9 | 882 | |||
AK166507.1 | - | 9 | 881 | |||
AK167209.1 | - | 9 | 881 | |||
AK167442.1 | - | 9 | 884 | |||
AK168710.1 | - | 9 | 863 | |||
AK185191.1 | - | 3 | 192 | |||
BC007467.1 | - | 8 | 819 | 3497714 | ||
D87115.1 | - | 9 | 893 | |||
X93150.1 | - | 7 | 615 |