Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008928.4 | NP_032954.1 | 3 | 318 | ||
GenBank mRNA | AK008141.1 | - | 3 | 321 | ||
AK011002.1 | - | 3 | 319 | |||
AK053378.1 | - | 3 | 319 | |||
AK150067.1 | - | 3 | 319 | |||
AK159379.1 | - | 3 | 317 | |||
AK166255.1 | - | 3 | 320 | |||
AK166507.1 | - | 3 | 319 | |||
AK167209.1 | - | 3 | 319 | |||
AK167442.1 | - | 3 | 322 | |||
AK168710.1 | - | 3 | 301 | |||
BC007467.1 | - | 3 | 309 | 3497714 | ||
D87115.1 | - | 3 | 331 | |||
X93150.1 | - | 1 | 53 |