Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168508.1 | NP_001161980.1 | 7 | 986 | ||
NM_001168513.1 | NP_001161985.1 | 7 | 587 | |||
NM_001168514.1 | NP_001161986.1 | 8 | 1031 | |||
NM_011951.3 | NP_036081.1 | 7 | 986 | |||
GenBank mRNA | AF128892.1 | - | 7 | 884 | ||
AF195850.1 | - | 4 | 309 | |||
AK133684.1 | - | 7 | 899 | |||
AK151348.1 | - | 7 | 588 | |||
AK153025.1 | - | 7 | 588 | |||
AK219197.1 | - | 1 | 43 | |||
CX568915.1 | - | 6 | 380 | |||
D83073.1 | - | 7 | 817 | |||
U10871.1 | - | 7 | 884 |