Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172300.3 | NP_758504.3 | 6 | 972 | ||
GenBank mRNA | AK005524.1 | - | 4 | 523 | ||
AK077793.1 | - | 6 | 971 | |||
AK149770.1 | - | 6 | 972 | |||
AK152529.1 | - | 6 | 686 | |||
AK153328.1 | - | 6 | 703 | |||
AK168942.1 | - | 6 | 665 | |||
BC027266.1 | - | 6 | 597 | |||
BC054412.1 | - | 6 | 675 | 2646768 |