Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf21b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081166.2 | NP_001074635.2 | 2, 3 | 526 | ||
GenBank mRNA | AK042863.1 | - | 2, 3 | 117 | ||
AK046530.1 | - | 2, 3 | 527 | |||
BC067021.1 | - | 2, 3 | 170 | |||
BC157925.1 | - | 2, 3 | 148 | 9088098 | ||
BC158044.1 | - | 2, 3 | 148 | 9007348 | ||
BC172019.1 | - | 2, 3 | 148 | 9054305 | ||
BC172145.1 | - | 2, 3 | 148 | 9054815 |