Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025887.4 | NP_080163.1 | 6 | 1020 | ||
GenBank mRNA | AB232593.1 | - | 5 | 553 | ||
AK002631.1 | - | 6 | 775 | |||
AK008198.1 | - | 6 | 761 | |||
AK010495.1 | - | 6 | 865 | |||
AK076093.1 | - | 6 | 1000 | |||
AK081999.1 | - | 6 | 1021 | |||
AK150296.1 | - | 6 | 774 | |||
AK150346.1 | - | 6 | 774 | |||
AK150405.1 | - | 6 | 818 | |||
AK151066.1 | - | 6 | 774 | |||
AK153131.1 | - | 6 | 774 | |||
AK159706.1 | - | 6 | 774 | |||
BC004842.1 | - | 6 | 881 | 2649317 | ||
BC034370.1 | - | 6 | 763 | 3709729 | ||
BC096481.1 | - | 7 | 954 | 5322132 |