Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf2bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178757.3 | NP_848872.2 | 1 | 2074 | ||
GenBank mRNA | AK031834.1 | - | 1 | 2082 | ||
AK032010.1 | - | 1 | 2019 | |||
AK083013.1 | - | 1 | 1715 | |||
AK087457.1 | - | 1 | 2087 | |||
BC019164.1 | - | 1 | 1755 | 3962494 | ||
BC036162.1 | - | 1 | 2074 | 4984359 |