Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163182.1 | NP_001156654.1 | 5, 6 | 650 | ||
NM_001163183.1 | NP_001156655.1 | 4, 5 | 503 | |||
NM_001163184.1 | NP_001156656.1 | 4, 5 | 503 | |||
NM_001163185.1 | NP_001156657.1 | 4, 5 | 493 | |||
NM_001163186.1 | NP_001156658.1 | 3, 4 | 494 | |||
NM_001163187.1 | NP_001156659.1 | 3, 4 | 494 | |||
NM_013746.3 | NP_038774.1 | 5, 6 | 650 | |||
GenBank mRNA | AF000272.1 | - | 5, 6 | 621 | ||
AF071000.1 | - | 3, 4 | 482 | |||
AF100613.1 | - | 3, 4 | 482 | |||
AF101053.1 | - | 5, 6 | 621 | |||
AK005102.1 | - | 4, 5 | 458 | |||
AK141634.1 | - | 461 | ||||
AK158172.1 | - | 4, 5 | 494 | |||
AK164683.1 | - | 4, 5 | 458 | |||
BC024756.1 | - | 5, 6 | 611 | 5369461 | ||
CX201870.1 | - | 4, 5 | 455 |