Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138586.3 | NP_613052.1 | 2 | 277 | ||
GenBank mRNA | AK006475.1 | - | 2 | 230 | ||
AK007372.1 | - | 2 | 266 | |||
AK137780.1 | - | 240 | ||||
AK155228.1 | - | 2 | 229 | |||
BB856072.1 | - | 2 | 279 | |||
BC034358.1 | - | 2 | 223 | 2651418 | ||
BC092393.1 | - | 2 | 218 | |||
BY196593.1 | - | 2 | 275 | |||
BY198385.1 | - | 2 | 275 |