Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013877.3 | NP_038905.1 | 5 | 610 | ||
GenBank mRNA | AB079605.1 | - | 4 | 426 | ||
AF169161.1 | - | 5 | 571 | |||
AK044544.1 | - | 5 | 611 | |||
AK087416.1 | - | 5 | 471 | |||
BC018253.1 | - | 5 | 633 | 4504283 |