Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slamf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030710.2 | NP_109635.1 | 3 | 633 | ||
GenBank mRNA | AB196808.1 | - | 3 | 462 | ||
AB196809.1 | - | 3 | 462 | |||
AB196810.1 | - | 3 | 462 | |||
AB196811.1 | - | 3 | 462 | |||
AB196836.1 | - | 3 | 462 | |||
AF248635.1 | - | 3 | 633 | |||
AF248636.1 | - | 3 | 655 | |||
BC030031.1 | - | 3 | 614 | 1225313 | ||
EU591721.1 | - | 3 | 462 |