Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198035.1 | NP_932152.1 | 2 | 2080 | ||
GenBank mRNA | AK135409.1 | - | 2 | 2073 | ||
AK155700.1 | - | 2 | 2057 | |||
AK172936.1 | - | 1 | 1833 | |||
BC058661.1 | - | 2 | 2080 | 6409441 | ||
BC060725.1 | - | 1 | 1689 | |||
BC079569.1 | - | 2 | 1976 | 30536951 |