Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iffo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028273.1 | - | 8, 9 | 1599 | ||
AK031129.1 | - | 7, 8 | 1823 | |||
AK044709.1 | - | 8, 9 | 1766 | |||
AK044728.1 | - | 9, 10 | 1595 | |||
AK053568.1 | - | 6, 7 | 2986 | |||
AK163571.1 | - | 8, 9 | 1595 | |||
AK169554.1 | - | 7, 8 | 1642 | |||
BC057601.1 | - | 9, 10 | 1044 | 6403474 |