Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iffo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028273.1 | - | 6 | 1328 | ||
AK031129.1 | - | 6 | 1308 | |||
AK044709.1 | - | 6 | 1495 | |||
AK044728.1 | - | 7 | 1327 | |||
AK053568.1 | - | 4 | 2715 | |||
AK163571.1 | - | 6 | 1327 | |||
AK169554.1 | - | 5 | 1374 | |||
AK184406.1 | - | 2 | 159 | |||
AK189508.1 | - | 2 | 159 | |||
BC057601.1 | - | 7 | 773 | 6403474 |