Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178035.4 | NP_828874.3 | 3 | 466 | ||
GenBank mRNA | AB087503.1 | - | 3 | 434 | ||
BC044804.1 | - | 3 | 324 | |||
BC132041.1 | - | 3 | 417 | 40130318 | ||
BC132067.1 | - | 3 | 417 | 40130344 |