Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181820.2 | NP_861541.2 | 12 | 1830 | ||
GenBank mRNA | AK050105.1 | - | 11 | 1725 | ||
AY236493.1 | - | 12 | 1793 | |||
AY263162.1 | - | 12 | 1801 | |||
BC022758.1 | - | 4 | 520 | |||
BC040466.1 | - | 12 | 1742 | |||
CN719678.1 | - | 1 | 24 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK132602.1 | - | 737 | Chr. - - [-] on Build |