Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181820.2 | NP_861541.2 | 15 | 2134 | ||
GenBank mRNA | AK050105.1 | - | 14 | 2029 | ||
AY236493.1 | - | 15 | 2097 | |||
AY263162.1 | - | 15 | 2105 | |||
BC022758.1 | - | 7 | 824 | |||
BC040466.1 | - | 15 | 2046 | |||
CN719678.1 | - | 4 | 328 |