Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194357.1 | NP_919338.2 | 8 | 860 | ||
GenBank mRNA | AK132627.1 | - | 3 | 212 | ||
BC048084.1 | - | 8 | 860 | 6312261 |