Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159421.1 | NP_001152893.1 | 4 | 712 | ||
NM_001159422.1 | NP_001152894.1 | 5 | 489 | |||
NM_198645.2 | NP_941047.1 | 5 | 838 | |||
GenBank mRNA | AK135450.1 | - | 5 | 418 | ||
BB686073.1 | - | 3 | 153 | |||
BC024406.1 | - | 5 | 402 | |||
BC024862.1 | - | 5 | 813 | 5356370 | ||
BC053690.1 | - | 5 | 390 | |||
CJ094333.1 | - | 4 | 293 |