Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001334.2 | NP_001001334.2 | 10 | 1454 | ||
GenBank mRNA | AK132882.1 | - | 10 | 1454 | ||
BC049634.1 | - | 6 | 638 | |||
BC061194.1 | - | 10 | 1451 |