Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009080.2 | NP_033106.1 | 4 | 524 | ||
GenBank mRNA | AK135286.1 | - | 4 | 495 | ||
AK152589.1 | - | 4 | 504 | |||
AK167245.1 | - | 4 | 503 | |||
AK169131.1 | - | 4 | 502 | |||
AK181061.1 | - | 118 | ||||
BC070397.1 | - | 4 | 588 | 5684553 | ||
BC099481.1 | - | 4 | 483 | 30847108 |
Gene Symbol: Gm15772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK146116.1 | - | 1 | 500 | ||
X80699.1 | - | 1 | 507 |