Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029645.3 | NP_083921.1 | 3 | 449 | ||
GenBank mRNA | AK008080.1 | - | 3 | 454 | ||
AK034326.1 | - | 3 | 449 | |||
BC021775.1 | - | 3 | 442 | 4158979 |