Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgprx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207540.3 | NP_997423.1 | 2 | 819 | ||
GenBank mRNA | AY152435.1 | - | 2 | 825 | ||
BC150669.1 | - | 2 | 769 | 9087580 |
Set Membership: |
G-protein coupled receptor |