Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008428.3 | NP_001008428.1 | 1 | 65 | ||
GenBank mRNA | AK135785.1 | - | 1 | 46 | ||
AK136200.1 | - | 1 | 43 | |||
BC085247.1 | - | 1 | 46 | 30477089 | ||
CJ076686.1 | - | 1 | 65 |