Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081069.1 | NP_001074538.1 | 13 | 965 | ||
GenBank mRNA | AK039314.1 | - | 12 | 1587 | ||
AK080707.1 | - | 13 | 1057 | |||
AK161551.1 | - | 9 | 1024 | |||
BC019741.1 | - | 2 | 297 | |||
BC049910.1 | - | 11 | 1669 |