Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016712.3 | NP_057921.1 | 4 | 422 | ||
GenBank mRNA | AF177174.1 | - | 3 | 341 | ||
AK144408.1 | - | 4 | 423 | |||
AK144535.1 | - | 4 | 423 | |||
BC068020.1 | - | 3 | 358 | 30291625 |