Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tollip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023764.3 | NP_076253.1 | 3 | 439 | ||
GenBank mRNA | AJ242971.1 | - | 3 | 443 | ||
AK015062.1 | - | 382 | ||||
AK016482.1 | - | 3 | 440 | |||
AK045422.1 | - | 3 | 423 | |||
AK049263.1 | - | 3 | 461 | |||
AK049394.1 | - | 3 | 385 | |||
AK078391.1 | - | 449 | ||||
AK161312.1 | - | 3 | 426 | |||
AK189903.1 | - | 2 | 213 | |||
BC053515.1 | - | 3 | 426 | |||
BC062139.1 | - | 3 | 420 | 6334946 |