Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tollip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023764.3 | NP_076253.1 | 6 | 892 | ||
GenBank mRNA | AJ242971.1 | - | 6 | 896 | ||
AK015062.1 | - | 835 | ||||
AK016482.1 | - | 6 | 893 | |||
AK045422.1 | - | 6 | 876 | |||
AK049263.1 | - | 6 | 914 | |||
AK049394.1 | - | 6 | 838 | |||
AK161312.1 | - | 6 | 879 | |||
BC053515.1 | - | 6 | 879 | |||
BC062139.1 | - | 6 | 873 | 6334946 |