Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019657.4 | NP_062631.1 | 10, 11 | 869 | ||
GenBank mRNA | AF064635.1 | - | 10, 11 | 848 | ||
AK012103.1 | - | 3, 4 | 283 | |||
AK078841.1 | - | 10, 11 | 870 | |||
AK081869.1 | - | 10, 11 | 863 | |||
AK082715.1 | - | 10, 11 | 863 | |||
AK088521.1 | - | 10, 11 | 863 | |||
AK150849.1 | - | 10, 11 | 864 | |||
AK152489.1 | - | 10, 11 | 864 | |||
AK166027.1 | - | 10, 11 | 863 | |||
AK167908.1 | - | 10, 11 | 863 | |||
AK188070.1 | - | 5, 6 | 346 | |||
AK212469.1 | - | 3, 4 | 259 | |||
BC010749.1 | - | 10, 11 | 844 | |||
BC037620.1 | - | 9, 10 | 786 | 5375831 | ||
BC090659.1 | - | 10, 11 | 839 | 30675153 |