Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111331.1 | NP_001104801.1 | 4 | 728 | ||
NM_001291005.1 | NP_001277934.1 | 5 | 550 | |||
NM_019789.4 | NP_062763.2 | 5 | 554 | |||
GenBank mRNA | AF184624.1 | - | 5 | 502 | ||
AF274050.1 | - | 4 | 414 | |||
AF287732.1 | - | 5 | 528 | |||
AF287733.1 | - | 5 | 532 | |||
AF300870.1 | - | 5 | 477 | |||
AK016721.1 | - | 5 | 716 | |||
AK165711.1 | - | 5 | 502 | |||
BC026980.1 | - | 5 | 491 | 5368268 | ||
BC047139.1 | - | 5 | 491 | 6389719 | ||
BC057329.1 | - | 5 | 473 | 6813819 | ||
DQ148499.1 | - | 4 | 414 | |||
DQ148500.1 | - | 4 | 728 |