Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Habp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019986.3 | NP_064370.2 | 6 | 985 | ||
GenBank mRNA | AF227684.1 | - | 6 | 928 | ||
AK016800.1 | - | 6 | 1028 | |||
AK144206.1 | - | 6 | 969 | |||
AK146525.1 | - | 6 | 958 | |||
BC082806.1 | - | 6 | 987 | |||
BC119460.2 | - | 5 | 481 | |||
BC148304.2 | - | 5 | 489 |