Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020044.3 | NP_064428.1 | 6 | 419 | ||
NM_022964.4 | NP_075253.2 | 6 | 419 | |||
GenBank mRNA | AF257136.1 | - | 5 | 363 | ||
AK134721.1 | - | 3 | 111 | |||
AK138953.1 | - | 6 | 378 | |||
AK143533.1 | - | 6 | 420 | |||
AK144261.1 | - | 6 | 358 | |||
AK162321.1 | - | 6 | 378 | |||
AY190024.1 | - | 6 | 384 | |||
BC005804.1 | - | 6 | 348 | 3154073 |