Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133788.2 | NP_598549.1 | 4 | 671 | ||
GenBank mRNA | AF209926.1 | - | 4 | 453 | ||
AK154135.1 | - | 4 | 603 | |||
AK154410.1 | - | 5 | 694 | |||
BC057132.1 | - | 5 | 690 | |||
BC110695.1 | - | 4 | 658 | 6842587 |