Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il21r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021887.2 | NP_068687.1 | 4 | 730 | ||
GenBank mRNA | AF254068.1 | - | 4 | 434 | ||
AF269134.1 | - | 3 | 292 | |||
AF279436.1 | - | 4 | 682 | |||
AF477982.1 | - | 3 | 308 | |||
AF477983.1 | - | 3 | 308 | |||
AF477984.1 | - | 3 | 308 | |||
AF477985.1 | - | 3 | 308 | |||
AK040073.1 | - | 3 | 628 | |||
AK137793.1 | - | 4 | 749 | |||
AK150824.1 | - | 4 | 691 | |||
AK171826.1 | - | 4 | 669 | |||
AK172032.1 | - | 4 | 702 | |||
BC057861.1 | - | 4 | 679 | 5315350 |