Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023065.3 | NP_075552.2 | 5 | 582 | ||
GenBank mRNA | AF290973.1 | - | 5 | 583 | ||
AF309649.1 | - | 5 | 550 | |||
AK077725.1 | - | 5 | 582 | |||
AK156914.1 | - | 5 | 583 | |||
BC054852.1 | - | 5 | 568 | 6528891 | ||
BE951500.1 | - | 3 | 412 |