Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Duoxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025777.2 | NP_080053.1 | 2 | 316 | ||
GenBank mRNA | AK008816.1 | - | 2 | 316 | ||
AK018569.1 | - | 2 | 318 |