Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3d21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162533.1 | NP_001156005.1 | 14 | 1534 | ||
NM_025856.3 | NP_080132.1 | 10 | 1198 | |||
GenBank mRNA | AK006490.1 | - | 1 | 319 | ||
AK076962.1 | - | 10 | 1163 | |||
AK155132.1 | - | 14 | 1455 | |||
BC019568.1 | - | 14 | 1451 | |||
BC025637.1 | - | 8 | 1195 | |||
BC039777.1 | - | 10 | 1388 | |||
BC053421.1 | - | 10 | 1159 | 6706860 |