Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aagab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025857.2 | NP_080133.1 | 4 | 429 | ||
GenBank mRNA | AK017714.1 | - | 4 | 398 | ||
AK159897.1 | - | 4 | 430 | |||
AK182339.1 | - | 3 | 184 | |||
AK186664.1 | - | 3 | 184 | |||
AK192186.1 | - | 3 | 185 | |||
AK209997.1 | - | 3 | 221 | |||
AK210563.1 | - | 3 | 184 | |||
AK214885.1 | - | 3 | 221 | |||
AK215257.1 | - | 3 | 221 | |||
BC027311.1 | - | 4 | 425 | 3964905 |