Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026405.3 | NP_080681.1 | 2 | 582 | ||
GenBank mRNA | AB232627.1 | - | 2 | 463 | ||
AK012701.1 | - | 2 | 594 | |||
AK078874.1 | - | 2 | 274 | |||
AK088661.1 | - | 2 | 583 | |||
AK151543.1 | - | 2 | 556 | |||
AK159102.1 | - | 2 | 609 | |||
AK159901.1 | - | 2 | 609 | |||
AK163620.1 | - | 2 | 533 | |||
AK180112.1 | - | 1 | 97 | |||
AK206948.1 | - | 2 | 292 | |||
AK216759.1 | - | 1 | 222 | |||
AY135650.1 | - | 2 | 487 | |||
BC016409.1 | - | 500 | 4456902 | |||
BC055945.1 | - | 508 | 4161169 |