Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rchy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271797.1 | NP_001258726.1 | 3 | 613 | ||
NM_026557.4 | NP_080833.1 | 4 | 733 | |||
GenBank mRNA | AF071222.1 | - | 4 | 405 | ||
AF276959.1 | - | 4 | 403 | |||
AF305423.1 | - | 4 | 389 | |||
AK015870.1 | - | 5 | 726 | |||
AK018364.1 | - | 4 | 400 | |||
AK018488.1 | - | 4 | 430 | |||
AK078397.1 | - | 4 | 517 | |||
BC023138.1 | - | 4 | 507 | 4008685 | ||
BC057143.1 | - | 4 | 733 | 5249543 | ||
DV662964.1 | - | 3 | 280 |