Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026637.3 | NP_080913.1 | 2, 3 | 320 | ||
GenBank mRNA | AK008719.1 | - | 2, 3 | 325 | ||
AK028830.1 | - | 322 | ||||
AK050049.1 | - | 2, 3 | 405 | |||
AK079465.1 | - | 2, 3 | 322 | |||
AK141474.1 | - | 370 | ||||
BC080818.1 | - | 2, 3 | 300 | 30286931 |