Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178615.3 | NP_848730.2 | 2 | 1011 | ||
GenBank mRNA | AJ557514.1 | - | 2 | 1011 | ||
AK004310.1 | - | 2 | 563 | |||
AK030386.1 | - | 2 | 967 | |||
AK035713.1 | - | 2 | 958 | |||
AK046637.1 | - | 2 | 959 | |||
AK047390.1 | - | 2 | 968 | |||
AK080819.1 | - | 2 | 958 | |||
BC030405.1 | - | 2 | 994 | |||
BC096024.1 | - | 2 | 958 | 6821755 | ||
BC138890.1 | - | 2 | 834 | 8861912 |