Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027117.3 | NP_081393.2 | 8, 9 | 1088 | ||
GenBank mRNA | AK009481.1 | - | 8, 9 | 1003 | ||
AK051418.1 | - | 8, 9 | 1015 | |||
AY513272.1 | - | 8, 9 | 1039 | |||
BC005581.1 | - | 8, 9 | 1012 | 3968615 |