Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027276.2 | NP_081552.2 | 17 | 1676 | ||
GenBank mRNA | AK013213.1 | - | 6 | 610 | ||
AK147938.1 | - | 17 | 1691 | |||
AK167965.1 | - | 17 | 1691 | |||
AK168978.1 | - | 17 | 1663 | |||
AK181696.1 | - | 3 | 258 | |||
AK184248.1 | - | 3 | 247 | |||
AK188605.1 | - | 3 | 252 | |||
AK198756.1 | - | 2 | 109 | |||
AK214776.1 | - | 3 | 205 | |||
BC005658.1 | - | 1 | 273 | |||
BC026606.1 | - | 17 | 1676 | 4976424 |