Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027276.2 | NP_081552.2 | 16 | 1584 | ||
GenBank mRNA | AK013213.1 | - | 5 | 518 | ||
AK147938.1 | - | 16 | 1599 | |||
AK167965.1 | - | 16 | 1599 | |||
AK168978.1 | - | 16 | 1571 | |||
AK181696.1 | - | 2 | 166 | |||
AK184248.1 | - | 2 | 155 | |||
AK188605.1 | - | 2 | 161 | |||
AK198756.1 | - | 1 | 17 | |||
AK201828.1 | - | 4 | 303 | |||
AK205790.1 | - | 2 | 141 | |||
AK214776.1 | - | 2 | 113 | |||
BC026606.1 | - | 16 | 1584 | 4976424 |