Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204931.1 | NP_001191860.1 | 3 | 375 | ||
NM_139292.2 | NP_647453.1 | 3 | 375 | |||
GenBank mRNA | AB039933.1 | - | 3 | 350 | ||
AK002562.1 | - | 3 | 373 | |||
AY562234.1 | - | 3 | 289 | |||
BC029741.1 | - | 3 | 362 | 4195889 |