Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2cbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027394.2 | NP_081670.1 | 8 | 961 | ||
GenBank mRNA | AK011446.1 | - | 8 | 978 | ||
AK049229.1 | - | 8 | 962 | |||
BC115686.1 | - | 8 | 930 | 40104268 | ||
BC115687.1 | - | 8 | 929 | 40104269 |