Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039081.2 | NP_001034170.1 | 8 | 842 | ||
NM_001286221.1 | NP_001273150.1 | 10 | 973 | |||
NM_001286222.1 | NP_001273151.1 | 10 | 1027 | |||
NM_001286223.1 | NP_001273152.1 | 8 | 823 | |||
NM_027696.3 | NP_081972.2 | 10 | 1030 | |||
GenBank mRNA | AK016915.1 | - | 2 | 272 | ||
AK129405.1 | - | 10 | 950 | |||
AY769435.1 | - | 9 | 822 | |||
AY769436.1 | - | 8 | 745 | |||
AY769437.1 | - | 8 | 795 | |||
AY769438.1 | - | 10 | 895 | |||
BC130227.1 | - | 8 | 794 | 40141863 | ||
BQ562973.1 | - | 5 | 420 | |||
CN538186.1 | - | 5 | 443 | |||
CN701986.1 | - | 2 | 196 |